Gene Transcripts
- Gene Symbol
- BRSK1
- Entrez Gene ID
- 84446
- Gene Name
- BR serine/threonine kinase 1
- Gene Aliases
- hSAD1
- Chromosome Location
- Chr. 19: 55284101 - 55312562 on Build GRCh38
- UniGene ID
- Hs.182081
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032430.1 |
NP_115806.1
|
10, 11 |
1314 |
|
XM_005259327.3 |
XP_005259384.1
|
10, 11 |
1383 |
|
XM_011527395.2 |
XP_011525697.2
|
8, 9 |
748 |
|
GenBank mRNA |
AB058714.1 |
- |
9, 10 |
849 |
|
AF479826.1 |
- |
12, 13 |
1212 |
|
AF479827.1 |
- |
10, 11 |
1314 |
|
AK124999.1 |
- |
2, 3 |
283 |
|
AL834275.1 |
- |
10, 11 |
937 |
|
AY458602.1 |
- |
10, 11 |
1086 |
|
AY505124.1 |
- |
10, 11 |
1037 |
|
BC016681.1 |
- |
2, 3 |
348 |
4341838 |
View Full Product Details