Gene Transcripts

Gene Symbol
B4galnt3
Entrez Gene ID
330406
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 3
Gene Aliases
AB114826, B4GalNAcT3, C330047A21
Chromosome Location
Chr. 6: 120200753 - 120295005 on Build GRCm38
UniGene ID
Mm.321840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198884.1 NP_942585.1 14 1352
GenBank mRNA AB114826.1 - 14 1352
AK143898.1 - 14 1266
BC139798.1 - 14 1366 40060840

View Full Product Details