Gene Transcripts

Gene Symbol
Rab40b
Entrez Gene ID
217371
Gene Name
Rab40B, member RAS oncogene family
Gene Aliases
SEC4L, rar
Chromosome Location
Chr. 11: 121356121 - 121388251 on Build GRCm38
UniGene ID
Mm.281639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139147.3 NP_631886.2 6 1031
GenBank mRNA AF425643.1 - 6 919
BC096601.1 - 6 1025 4974190

View Full Product Details