Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144902.3 | NP_659151.1 | 7 | 1048 | |
GenBank mRNA | AK029398.1 | - | 7 | 1420 | |
AK041195.1 | - | 8 | 1168 | ||
AK049484.1 | - | 7 | 1048 | ||
AK161317.1 | - | 7 | 1006 | ||
BC024110.1 | - | 7 | 1044 | 5150304 |