Gene Transcripts
- Gene Symbol
- Vps18
- Entrez Gene ID
- 228545
- Gene Name
- vacuolar protein sorting 18 (yeast)
- Gene Aliases
- 9930024E13Rik
- Chromosome Location
- Chr. 2: 119288742 - 119298453 on Build GRCm38
- UniGene ID
- Mm.294761
- Species
- Mus musculus
- Gene Symbol
- Atp6v0c
- Entrez Gene ID
- 11984
- Gene Name
- ATPase, H+ transporting, lysosomal V0 subunit C
- Gene Aliases
- Atp6c, Atp6c2, Atp6l, Atpl, Atpl-rs1, PL16, VATL, Vma3
- Chromosome Location
- Chr. 17: 24163866 - 24169429 on Build GRCm38
- UniGene ID
- Mm.294761
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK189660.1 |
- |
|
111 |
|
View Full Product Details