Gene Transcripts

Gene Symbol
Adamts2
Entrez Gene ID
216725
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 2
Gene Aliases
ADAM-TS2, ADAMTS-2, ADAMTS-3, PCINP, hPCPNI, mKIAA4060, pNPI
Chromosome Location
Chr. 11: 50602085 - 50807573 on Build GRCm38
UniGene ID
Mm.110597
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277305.1 NP_001264234.1 2 545
NM_175643.3 NP_783574.1 2 545
GenBank mRNA AK220464.1 - 1 287
BC046456.1 - 2 545 3965027

View Full Product Details