Gene Transcripts

Gene Symbol
B3gnt4
Entrez Gene ID
231727
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Gene Aliases
1010001G17Rik, BGnT-4
Chromosome Location
Chr. 5: 123508289 - 123512099 on Build GRCm38
UniGene ID
Mm.484068
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198611.2 NP_941013.2 1 1144
GenBank mRNA AY037786.1 - 1 1145
BC114987.1 - 1 1099 40056240
BC115755.1 - 1 1099 40056241

View Full Product Details