Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145806.1 | NP_001139278.1 | 3 | 419 | |
NM_130890.2 | NP_570960.2 | 3 | 419 | ||
GenBank mRNA | AB050202.1 | - | 3 | 415 | |
AB050203.1 | - | 3 | 415 | ||
AB061518.1 | - | 3 | 365 | ||
AB061519.1 | - | 3 | 365 | ||
AK132449.1 | - | 3 | 420 | ||
BC125504.1 | - | 3 | 391 | 40130019 | |
BC125508.1 | - | 3 | 391 | 40130023 | |
BC145055.1 | - | 3 | 391 |