Gene Transcripts

Gene Symbol
C8b
Entrez Gene ID
110382
Gene Name
complement component 8, beta polypeptide
Gene Aliases
4930439B20Rik, AI595927
Chromosome Location
Chr. 4: 104764576 - 104804549 on Build GRCm38
UniGene ID
Mm.87072
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133882.2 NP_598643.1 3 436
GenBank mRNA AB077304.1 - 3 358
AB077306.1 - 3 358
AK050313.1 - 3 437
BC022129.1 - 1 67
BC096382.1 - 3 410 5102825

View Full Product Details