Gene Transcripts

Gene Symbol
Acat1
Entrez Gene ID
110446
Gene Name
acetyl-Coenzyme A acetyltransferase 1
Gene Aliases
6330585C21Rik, Acat
Chromosome Location
Chr. 9: 53580522 - 53610350 on Build GRCm38
UniGene ID
Mm.293233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144784.3 NP_659033.1 7 701
GenBank mRNA AJ510150.1 - 7 693
AK032097.1 - 7 672
AK075593.1 - 7 674
AK081715.1 - 7 701
AK146241.1 - 7 656
AK163402.1 - 7 705
AK169771.1 - 7 694
BC024429.1 - 7 704
BC024763.1 - 7 673 5365469
BC029345.1 - 4 362

View Full Product Details