Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172448.3 | NP_766036.2 | 9 | 2195 | |
GenBank mRNA | AK028750.1 | - | 8 | 1554 | |
BC029717.1 | - | 1 | 571 | ||
BC037465.1 | - | 1 | 635 | ||
BC075707.1 | - | 7 | 1331 | ||
BC131957.1 | - | 7 | 1313 | 40130234 |