Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001105529.1 | NP_001098999.1 | 2 | 339 | |
NM_006095.2 | NP_006086.1 | 2 | 339 | ||
XM_005248043.2 | XP_005248100.1 | 2 | 334 | ||
XM_011513615.1 | XP_011511917.1 | 2 | 334 | ||
XM_011513616.2 | XP_011511918.1 | 2 | 368 | ||
XM_011513618.1 | XP_011511920.1 | 2 | 334 | ||
XM_017007645.1 | XP_016863134.1 | 2 | 368 | ||
XM_017007646.1 | XP_016863135.1 | 2 | 315 | ||
XM_017007647.1 | XP_016863136.1 | 2 | 313 | ||
GenBank mRNA | AB013452.1 | - | 2 | 120 | |
AB209687.1 | - | 2 | 192 | ||
AF067820.1 | - | 2 | 107 | ||
BC020943.1 | - | 2 | 327 | ||
BC109317.1 | - | 2 | 101 | 40036536 | |
BC109318.1 | - | 2 | 107 | 40036538 | |
BP232466.1 | - | 2 | 339 |