Gene Transcripts

Gene Symbol
Plet1
Entrez Gene ID
76509
Gene Name
placenta expressed transcript 1
Gene Aliases
0610037B23Rik, 1600029D21Rik, AA407923, AU043048
Chromosome Location
Chr. 9: 50494525 - 50505639 on Build GRCm38
UniGene ID
Mm.29959
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029639.2 NP_083915.2 4 1028
GenBank mRNA AK005558.1 - 4 1031
BC017624.1 - 4 1012 4487871
BC022950.1 - 4 1008 5148408

View Full Product Details