Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033581.3 | NP_291059.1 | 1 | 360 | |
GenBank mRNA | AK089310.1 | - | 1 | 406 | |
AY013811.1 | - | 1 | 222 | ||
BC059175.1 | - | 1 | 360 | 5704674 | |
BC098457.1 | - | 1 | 354 | 30653042 |