Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102468.1 | NP_001095938.1 | 4 | 581 | |
NM_138304.2 | NP_612177.1 | 5 | 722 | ||
GenBank mRNA | AY061807.1 | - | 5 | 722 | |
BC013541.1 | - | 5 | 700 | 4241711 | |
BC023475.1 | - | 5 | 687 | 5042406 |