Gene Transcripts

Gene Symbol
Afap1
Entrez Gene ID
70292
Gene Name
actin filament associated protein 1
Gene Aliases
2600003E23Rik, 9630044L16Rik, AI848729, Afap, mKIAA3018
Chromosome Location
Chr. 5: 35893317 - 36003923 on Build GRCm38
UniGene ID
Mm.125167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027373.2 NP_081649.1 12, 13 1759
GenBank mRNA AK129483.1 - 12, 13 1686
AK147706.1 - 12, 13 1651
AK148000.1 - 12, 13 1614
AK161385.1 - 12, 13 1710
BC050814.1 - 12, 13 1759 6773872

View Full Product Details