Gene Transcripts

Gene Symbol
Htra2
Entrez Gene ID
64704
Gene Name
HtrA serine peptidase 2
Gene Aliases
AI481710, Omi, Prss25, mnd2
Chromosome Location
Chr. 6: 83051266 - 83054571 on Build GRCm38
UniGene ID
Mm.21880
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019752.3 NP_062726.3 1 587
GenBank mRNA AF164513.1 - 1 466
AF175324.1 - 1 483
AK011536.1 - 1290
AK146338.1 - 1 478
AK159188.1 - 1 407
BC033276.1 - 1 447
BC049880.1 - 1 587 5321861

View Full Product Details