Gene Transcripts

Gene Symbol
Bin3
Entrez Gene ID
57784
Gene Name
bridging integrator 3
Gene Aliases
1700015F07Rik
Chromosome Location
Chr. 14: 70100090 - 70138206 on Build GRCm38
UniGene ID
Mm.24186
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021328.3 NP_067303.1 6, 7 380
GenBank mRNA AF244361.1 - 6, 7 360
AF271733.1 - 6, 7 423
AK005992.1 - 6, 7 389
AK040685.1 - 6, 7 380
AK170915.1 - 6, 7 421
AK210433.1 - 6, 7 361
BC026543.1 - 6, 7 365 5040094

View Full Product Details