Gene Transcripts

Gene Symbol
Anapc5
Entrez Gene ID
59008
Gene Name
anaphase-promoting complex subunit 5
Gene Aliases
2510006G12Rik, AA408751, AA536819, AA986414, APC5, Anpc5
Chromosome Location
Chr. 5: 122787459 - 122821342 on Build GRCm38
UniGene ID
Mm.332667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042491.2 NP_001035956.1 3 539
NM_001289517.1 NP_001276446.1 3 539
NM_001289518.1 NP_001276447.1 3 539
NM_001289519.1 NP_001276448.1 3 539
NM_001289520.1 NP_001276449.1 3 539
NM_021505.3 NP_067480.2 3 539
GenBank mRNA AB041593.1 - 3 482
AK013978.1 - 3 463
AK031432.1 - 3 464
AK049719.1 - 3 493
AK075969.1 - 3 478
AK088327.1 - 3 502
AK147951.1 - 3 478
AK148679.1 - 3 494
AK151314.1 - 3 485
AK151905.1 - 3 485
AK152528.1 - 3 455
AK152981.1 - 3 485
AK153373.1 - 3 485
AK153422.1 - 3 485
AK159204.1 - 3 485
AK159713.1 - 3 478
AK159916.1 - 3 478
AK167561.1 - 3 478
AK168135.1 - 3 463
AK168737.1 - 3 479
AK168814.1 - 3 478
AK169196.1 - 3 502
AK186196.1 - 3 217
AK192754.1 - 3 235
AK194108.1 - 3 204
AK197886.1 - 3 211
AK203216.1 - 2 188
AK215079.1 - 3 212
AK217317.1 - 3 217
BC010339.1 - 3 455 2648173
BC046566.1 - 3 518 2650790
BC046804.1 - 3 476 6404923

View Full Product Details