Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301643.1 | NP_001288572.1 | 7 | 503 | |
NM_001301644.1 | NP_001288573.1 | 7 | 472 | ||
NM_019640.5 | NP_062614.1 | 7 | 503 | ||
GenBank mRNA | AK028235.1 | - | 7 | 438 | |
BC034676.1 | - | 7 | 393 | 4481685 | |
CK619028.1 | - | 7 | 386 | ||
U46934.1 | - | 7 | 405 |