Gene Transcripts

Gene Symbol
Akap8
Entrez Gene ID
56399
Gene Name
A kinase (PRKA) anchor protein 8
Gene Aliases
1200016A02Rik, AA673585, AKAP-8, AKAP95, AU015639
Chromosome Location
Chr. 17: 32303672 - 32321327 on Build GRCm38
UniGene ID
Mm.328945
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB028920.1 - 4 295
AK004801.1 - 4 301
AK030964.1 - 1 935
AK089092.1 - 4 308
AK143300.1 - 2 1683
BC042602.1 - 4 291
BC060170.1 - 4 286
BC060209.1 - 4 392
BC078448.1 - 4 279
BC125521.1 - 4 291 40130036
BC137746.1 - 4 291 8860765

View Full Product Details