Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019938.3 | NP_064322.1 | 3 | 200 | |
GenBank mRNA | AB029919.1 | - | 3 | 200 | |
AK133053.1 | - | 2 | 256 | ||
BC138245.1 | - | 3 | 152 | 8861264 | |
BC138246.1 | - | 3 | 152 | 8861265 |