Gene Transcripts

Gene Symbol
Mocs1
Entrez Gene ID
56738
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
3110045D15Rik, AI043170, AW536397
Chromosome Location
Chr. 17: 49428364 - 49455435 on Build GRCm38
UniGene ID
Mm.22256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020042.2 NP_064426.2 11 1721
NM_028464.1 NP_082740.1 11 1736
GenBank mRNA AF214016.1 - 5 911
AK157231.1 - 11 1677
BC027444.1 - 3 647
BC034066.1 - 7 1085
BC049914.1 - 9 1364
BC147411.1 - 2 565
BC147412.1 - 2 550
BC172067.1 - 2 550

View Full Product Details