Gene Transcripts

Gene Symbol
ADRM1
Entrez Gene ID
11047
Gene Name
adhesion regulating molecule 1
Gene Aliases
ARM-1, ARM1, GP110
Chromosome Location
Chr. 20: 62302056 - 62308862 on Build GRCh38
UniGene ID
Hs.90107
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281437.1 NP_001268366.1 2 292
NM_001281438.1 NP_001268367.1 2 296
NM_007002.3 NP_008933.2 2 296
NM_175573.2 NP_783163.1 2 292
XM_005260257.1 XP_005260314.1 3 383
XM_011528503.1 XP_011526805.1 2 1131
XM_017027602.1 XP_016883091.1 2 1131
XM_017027603.1 XP_016883092.1 3 383
XM_017027604.1 XP_016883093.1 2 1127
XM_017027605.1 XP_016883094.1 2 1131
XM_017027606.1 XP_016883095.1 2 1131
XM_017027607.1 XP_016883096.1 2 1127
GenBank mRNA AK297567.1 - 2 190
AK307459.1 - 226
AL558522.3 - 2 228
BC017245.2 - 2 192 4890786
BG337785.1 - 2 252
CR749213.1 - 1 234
D64154.1 - 2 187
HY029018.1 - 3 389
HY178253.1 - 2 242

View Full Product Details