Gene Transcripts
- Gene Symbol
- C3ar1
- Entrez Gene ID
- 84007
- Gene Name
- complement component 3a receptor 1
- Gene Aliases
- Chromosome Location
- Chr. 4: 155681767 - 155691114 on Build Rnor_6.0
- UniGene ID
- Rn.226511
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032060.1 |
NP_114449.1
|
2 |
1680 |
|
GenBank mRNA |
U86379.1 |
- |
2 |
1680 |
|
View Full Product Details