Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007033.4 | NP_008964.3 | 5 | 543 | |
XM_005244713.4 | XP_005244770.2 | 5 | 1364 | ||
XM_006710306.2 | XP_006710369.1 | 6 | 625 | ||
XM_011540542.1 | XP_011538844.1 | 6 | 563 | ||
XM_011540543.1 | XP_011538845.1 | 5 | 411 | ||
XM_017000131.1 | XP_016855620.1 | 5 | 1894 | ||
XM_017000132.1 | XP_016855621.1 | 6 | 653 | ||
XM_017000133.1 | XP_016855622.1 | 6 | 736 | ||
XM_017000134.1 | XP_016855623.1 | 7 | 650 | ||
XM_017000135.1 | XP_016855624.1 | 5 | 650 | ||
GenBank mRNA | AF091071.1 | - | 5 | 428 | |
AF157324.1 | - | 5 | 482 | ||
AJ001421.1 | - | 4 | 352 | ||
AK095750.1 | - | 4 | 2138 | ||
AK289658.1 | - | 5 | 477 | ||
BC004965.1 | - | 5 | 478 | 3546970 | |
BT007224.1 | - | 4 | 352 | ||
CR456806.1 | - | 4 | 352 |