Gene Transcripts

Gene Symbol
Serinc1
Entrez Gene ID
294421
Gene Name
serine incorporator 1
Gene Aliases
LRRGT00191, Tde1l, Tde2
Chromosome Location
Chr. 20: 38966016 - 38985056 on Build Rnor_6.0
UniGene ID
Rn.2078
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182951.1 NP_891996.1 3 291
GenBank mRNA AY339894.1 - 3 291
AY539942.1 - 3939
BC088852.1 - 3 302 7315523
DQ103708.1 - 3 271

View Full Product Details