Gene Transcripts

Gene Symbol
ADAMTS5
Entrez Gene ID
11096
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 5
Gene Aliases
ADAM-TS 11, ADAM-TS 5, ADAM-TS5, ADAMTS-11, ADAMTS-5, ADAMTS11, ADMP-2
Chromosome Location
Chr. 21: 26917912 - 26967120 on Build GRCh38
UniGene ID
Hs.58324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007038.4 NP_008969.2 8 4123
XM_011529448.2 XP_011527750.1 7 3955
GenBank mRNA AF142099.1 - 8 3516

View Full Product Details