Gene Transcripts

Gene Symbol
Bzw2
Entrez Gene ID
171439
Gene Name
basic leucine zipper and W2 domains 2
Gene Aliases
Hfb2
Chromosome Location
Chr. 6: 55586754 - 55647650 on Build Rnor_6.0
UniGene ID
Rn.13196
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134402.3 NP_599229.1 3 275
GenBank mRNA AF031483.1 - 3 303
BC063149.1 - 3 279 6890706
FQ225034.1 - 3 251
FQ228189.1 - 3 251

View Full Product Details