Gene Transcripts

Gene Symbol
Atp12a
Entrez Gene ID
171028
Gene Name
ATPase, H+/K+ transporting, nongastric, alpha polypeptide
Gene Aliases
Chromosome Location
Chr. 15: 36565495 - 36590171 on Build Rnor_6.0
UniGene ID
Rn.9858
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301664.1 NP_001288593.1 1 521
NM_133517.2 NP_598201.2 2 380
GenBank mRNA M90398.1 - 2 304
U94912.1 - 2 376
U94913.1 - 1 520

View Full Product Details