Gene Transcripts

Gene Symbol
Atp1b3
Entrez Gene ID
25390
Gene Name
ATPase, Na+/K+ transporting, beta 3 polypeptide
Gene Aliases
NKAB3S
Chromosome Location
Chr. 8: 104190032 - 104221342 on Build Rnor_6.0
UniGene ID
Rn.5041
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012913.1 NP_037045.1 3 412
GenBank mRNA BC061719.1 - 3 415
D84450.1 - 3 412

View Full Product Details