Gene Transcripts

Gene Symbol
Btg2
Entrez Gene ID
29619
Gene Name
BTG family, member 2
Gene Aliases
Agl, An, Pc3, Tis21, an-1
Chromosome Location
Chr. 13: 50913185 - 50916944 on Build Rnor_6.0
UniGene ID
Rn.27923
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017259.1 NP_058955.1 2 2475
GenBank mRNA BC072493.1 - 2 2434
M60921.1 - 2 2475

View Full Product Details