Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012497.1 | NP_036629.1 | 5 | 533 | |
GenBank mRNA | BC099749.1 | - | 5 | 519 | |
FQ213506.1 | - | 5 | 536 | ||
FQ214420.1 | - | 5 | 559 | ||
FQ228893.1 | - | 5 | 533 | ||
X06984.1 | - | 5 | 533 |