Gene Transcripts

Gene Symbol
Bgn
Entrez Gene ID
25181
Gene Name
biglycan
Gene Aliases
BSPG1
Chromosome Location
Chr. X: 157319042 - 157331204 on Build Rnor_6.0
UniGene ID
Rn.783
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017087.1 NP_058783.1 8 1428
GenBank mRNA AY724513.1 - 8 1433
BC072480.1 - 8 1438 7104041
U17834.1 - 8 1428

View Full Product Details