Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014436.2 | NP_001014436.1 | 2 | 209 | |
NM_001122956.1 | NP_001116428.1 | 2 | 209 | ||
NM_001284315.1 | NP_001271244.1 | 2 | 209 | ||
NM_014063.6 | NP_054782.2 | 2 | 209 | ||
XM_011515325.1 | XP_011513627.1 | 2 | 180 | ||
GenBank mRNA | AF077353.1 | - | 2 | 149 | |
AF151364.1 | - | 2 | 121 | ||
AF197060.1 | - | 2 | 111 | ||
AF218020.3 | - | 2 | 143 | ||
AF250287.1 | - | 2 | 111 | ||
AK027367.1 | - | 2 | 138 | ||
AK293151.1 | - | 2 | 147 | ||
AK293279.1 | - | 2 | 176 | ||
AK293340.1 | - | 2 | 141 | ||
AK300630.1 | - | 2 | 129 | ||
AK302037.1 | - | 2 | 193 | ||
BC011677.2 | - | 2 | 132 | 4299867 | |
BC031687.1 | - | 2 | 117 | 5168099 | |
CR457167.1 | - | 2 | 111 | ||
DC404852.1 | - | 2 | 209 |