Gene Transcripts

Gene Symbol
ADAMTS2
Entrez Gene ID
9509
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 2
Gene Aliases
ADAM-TS2, ADAMTS-2, ADAMTS-3, NPI, PC I-NP, PCI-NP, PCINP, PCPNI, PNPI
Chromosome Location
Chr. 5: 179110851 - 179345430 on Build GRCh38
UniGene ID
Hs.23871
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014244.4 NP_055059.2 3 665
NM_021599.3 NP_067610.1 3 665
XM_017010063.1 XP_016865552.1 2 101
GenBank mRNA AJ003125.1 - 3 563
AK308213.1 - 3 668

View Full Product Details