Gene Transcripts

Gene Symbol
RBPJL
Entrez Gene ID
11317
Gene Name
recombination signal binding protein for immunoglobulin kappa J region like
Gene Aliases
RBPL, RBPSUHL, SUHL
Chromosome Location
Chr. 20: 45306836 - 45319677 on Build GRCh38
UniGene ID
Hs.248217
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281448.1 NP_001268377.1 9, 10 1093
NM_001281449.1 NP_001268378.1 9, 10 1093
NM_014276.3 NP_055091.2 9, 10 1093
XM_011528521.2 XP_011526823.1 9, 10 1100
GenBank mRNA AB024964.1 - 9, 10 1013
AB026048.1 - 9, 10 1013
AB027710.1 - 9, 10 1013

View Full Product Details