Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286414.2 | NP_001273343.1 | 3 | 539 | |
NM_014444.4 | NP_055259.2 | 3 | 539 | ||
XM_011521454.2 | XP_011519756.1 | 3 | 539 | ||
XM_011521455.2 | XP_011519757.1 | 3 | 437 | ||
XM_017022078.1 | XP_016877567.1 | 3 | 539 | ||
GenBank mRNA | AJ249677.1 | - | 3 | 245 | |
AK026551.1 | - | 3 | 526 | ||
AK027703.1 | - | 3 | 497 | ||
BC009870.2 | - | 3 | 464 | 3940649 | |
BC012801.2 | - | 3 | 483 | 2821891 | |
DA396753.1 | - | 3 | 539 |