Gene Transcripts

Gene Symbol
OR9I1
Entrez Gene ID
219954
Gene Name
olfactory receptor family 9 subfamily I member 1
Gene Aliases
OR11-228
Chromosome Location
Chr. 11: 58118500 - 58119444 on Build GRCh38
UniGene ID
Hs.632122
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005211.1 NP_001005211.1 1 585

View Full Product Details