Gene Transcripts

Gene Symbol
ANKRD34B
Entrez Gene ID
340120
Gene Name
ankyrin repeat domain 34B
Gene Aliases
DP58
Chromosome Location
Chr. 5: 80556755 - 80570485 on Build GRCh38
UniGene ID
Hs.519404
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004441.2 NP_001004441.2 5 1836
XM_006714599.3 XP_006714662.1 5 1598
XM_011543366.2 XP_011541668.1 5 1640
XM_011543369.2 XP_011541671.1 3 1596
GenBank mRNA BC137441.1 - 5 1615
BC137442.1 - 5 1612 9021440
BC142957.1 - 4 1516 8860359
CR749535.1 - 5 1630

View Full Product Details