Gene Transcripts

Gene Symbol
PATE2
Entrez Gene ID
399967
Gene Name
prostate and testis expressed 2
Gene Aliases
C11orf38, PATE-M, UNQ3112
Chromosome Location
Chr. 11: 125776130 - 125778819 on Build GRCh38
UniGene ID
Hs.369185
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212555.2 NP_997720.1 4 579
GenBank mRNA AY358105.1 - 4 579
BC137190.1 - 4 570 9021187
BC137193.1 - 4 570 9021190
BC144526.1 - 3 441 9053053

View Full Product Details