Gene Transcripts

Gene Symbol
MT4
Entrez Gene ID
84560
Gene Name
metallothionein 4
Gene Aliases
MT-4, MT-IV, MTIV
Chromosome Location
Chr. 16: 56565049 - 56568957 on Build GRCh38
UniGene ID
Hs.567624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032935.2 NP_116324.1 1 99
GenBank mRNA BC113442.1 - 1 99
BC113444.1 - 1 99 8322496

View Full Product Details