Gene Transcripts
- Gene Symbol
- ZNF335
- Entrez Gene ID
- 63925
- Gene Name
- zinc finger protein 335
- Gene Aliases
- MCPH10, NIF-1, NIF1, NIF2
- Chromosome Location
- Chr. 20: 45948653 - 45972910 on Build GRCh38
- UniGene ID
- Hs.174193
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022095.3 |
NP_071378.1
|
24, 25 |
3800 |
|
XM_005260504.4 |
XP_005260561.1
|
24, 25 |
4513 |
|
XM_017028012.1 |
XP_016883501.1
|
23, 24 |
3571 |
|
GenBank mRNA |
AF395833.1 |
- |
24, 25 |
3777 |
|
AK026157.1 |
- |
16, 17 |
2315 |
|
AK074214.1 |
- |
9, 10 |
1435 |
|
AK122963.1 |
- |
7, 8 |
1991 |
|
AK296989.1 |
- |
21, 22 |
3371 |
|
BC041331.2 |
- |
21, 22 |
3015 |
|
BC142981.1 |
- |
21, 22 |
3015 |
|
View Full Product Details