Gene Transcripts

Gene Symbol
AK9
Entrez Gene ID
221264
Gene Name
adenylate kinase 9
Gene Aliases
AK 9, AKD1, AKD2, C6orf199, C6orf224, dJ70A9.1
Chromosome Location
Chr. 6: 109492855 - 109691253 on Build GRCh38
UniGene ID
Hs.205144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017010388.1 XP_016865877.1 31 5051
GenBank mRNA AK131244.1 - 5 1853
BC087860.1 - 5 913

View Full Product Details