Gene Transcripts

Gene Symbol
C10orf99
Entrez Gene ID
387695
Gene Name
chromosome 10 open reading frame 99
Gene Aliases
AP-57, CSBF, UNQ1833
Chromosome Location
Chr. 10: 84173764 - 84185294 on Build GRCh38
UniGene ID
Hs.298713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207373.2 NP_997256.1 3 470
XM_005269811.3 XP_005269868.1 3 334
GenBank mRNA AY358751.1 - 3 457

View Full Product Details