Gene Transcripts

Gene Symbol
C1qa
Entrez Gene ID
298566
Gene Name
complement component 1, q subcomponent, A chain
Gene Aliases
Chromosome Location
Chr. 5: 155261254 - 155264101 on Build Rnor_6.0
UniGene ID
Rn.105647
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008515.1 NP_001008515.1 3 948
GenBank mRNA BC086605.1 - 3 948

View Full Product Details