Gene Transcripts

Gene Symbol
MC4R
Entrez Gene ID
4160
Gene Name
melanocortin 4 receptor
Gene Aliases
Chromosome Location
Chr. 18: 60371331 - 60372768 on Build GRCh38
UniGene ID
Hs.532833
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005912.2 NP_005903.2 1 867
GenBank mRNA AK314130.1 - 1 867
BC069172.1 - 1 448 7216994
BC101802.1 - 1 508 8069308
BC111992.1 - 1 508 8327460

View Full Product Details