Gene Transcripts

Gene Symbol
CAMTA1
Entrez Gene ID
23261
Gene Name
calmodulin binding transcription activator 1
Gene Aliases
CANPMR
Chromosome Location
Chr. 1: 6785324 - 7769706 on Build GRCh38
UniGene ID
Hs.397705
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015215.3 NP_056030.1 23 6090
XM_011541083.2 XP_011539385.1 23 5944
XM_011541084.2 XP_011539386.1 24 5975
XM_011541085.2 XP_011539387.1 24 6016
XM_011541086.2 XP_011539388.1 22 5923
XM_011541087.2 XP_011539389.1 22 5872
XM_011541088.2 XP_011539390.1 22 5874
XM_011541089.2 XP_011539391.1 23 5605
XM_011541090.2 XP_011539392.1 24 5636
XM_017000773.1 XP_016856262.1 24 6696
XM_017000775.1 XP_016856264.1 22 5884
XM_017000776.1 XP_016856265.1 25 6076
XM_017000777.1 XP_016856266.1 22 5584
XM_017000778.1 XP_016856267.1 23 5615
XM_017000779.1 XP_016856268.1 23 5566
GenBank mRNA AB020640.3 - 23 6091
AF111804.1 - 6 1549
BC038183.2 - 1 886
BC051341.1 - 11 2743
BC151835.1 - 23 6091 8860115

View Full Product Details