Gene Transcripts
- Gene Symbol
- ARHGAP25
- Entrez Gene ID
- 9938
- Gene Name
- Rho GTPase activating protein 25
- Gene Aliases
- HEL-S-308, KAIA0053
- Chromosome Location
- Chr. 2: 68710505 - 68826833 on Build GRCh38
- UniGene ID
- Hs.531807
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007231.2 |
NP_001007232.2
|
10, 11 |
2146 |
|
NM_001166276.1 |
NP_001159748.1
|
9, 10 |
2086 |
|
NM_001166277.1 |
NP_001159749.1
|
9, 10 |
2026 |
|
NM_014882.2 |
NP_055697.1
|
9, 10 |
2083 |
|
XM_005264675.2 |
XP_005264732.1
|
10, 11 |
1850 |
|
XM_005264676.2 |
XP_005264733.1
|
9, 10 |
1734 |
|
XM_011533206.2 |
XP_011531508.1
|
10, 11 |
1894 |
|
XM_011533207.2 |
XP_011531509.1
|
10, 11 |
1860 |
|
XM_011533208.2 |
XP_011531510.1
|
11, 12 |
1911 |
|
XM_011533209.2 |
XP_011531511.1
|
7, 8 |
1451 |
|
XM_011533210.2 |
XP_011531512.1
|
5, 6 |
978 |
|
XM_017005423.1 |
XP_016860912.1
|
10, 11 |
1754 |
|
XM_017005424.1 |
XP_016860913.1
|
10, 11 |
1751 |
|
XM_017005425.1 |
XP_016860914.1
|
9, 10 |
1637 |
|
XM_017005426.1 |
XP_016860915.1
|
7, 8 |
2088 |
|
XM_017005427.1 |
XP_016860916.1
|
7, 8 |
2085 |
|
GenBank mRNA |
AK290396.1 |
- |
10, 11 |
1824 |
|
AK297056.1 |
- |
9, 10 |
1904 |
|
AK297157.1 |
- |
10, 11 |
1886 |
|
AK298669.1 |
- |
9, 10 |
1913 |
|
D29642.1 |
- |
9, 10 |
1896 |
|
EU794656.1 |
- |
9, 10 |
2083 |
|
View Full Product Details