Gene Transcripts
- Gene Symbol
- B3galnt2
- Entrez Gene ID
- 97884
- Gene Name
- UDP-GalNAc:betaGlcNAc beta 1,3-galactosaminyltransferase, polypeptide 2
- Gene Aliases
- A930105D20Rik, C80633, D230016N13Rik
- Chromosome Location
- Chr. 13: 13954474 - 13999098 on Build GRCm38
- UniGene ID
- Mm.21686
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178640.2 |
NP_848755.1
|
7, 8 |
989 |
|
GenBank mRNA |
AB116655.1 |
- |
7, 8 |
858 |
|
AK035259.1 |
- |
7, 8 |
990 |
|
AK041022.1 |
- |
7, 8 |
985 |
|
AK084275.1 |
- |
7, 8 |
989 |
|
AK085691.1 |
- |
7, 8 |
991 |
|
AK144880.1 |
- |
7, 8 |
820 |
|
AK151677.1 |
- |
7, 8 |
1039 |
|
AK153362.1 |
- |
7, 8 |
1039 |
|
AK167635.1 |
- |
7, 8 |
990 |
|
BC085110.1 |
- |
7, 8 |
998 |
30735904 |
View Full Product Details